Learn More
Zinc peroxide, approximately 50% ZnO{2}, Thermo Scientific Chemicals
Curing agent, rubber accelerator
Supplier: Thermo Scientific Chemicals 03963522
Description
Zinc peroxide is used as curing agent, rubber accelerator. has been applied in a variety of settings, from medicine to aesthetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc peroxide | |
1314-22-3 | |
100 g | |
MFCD00137469 | |
14,10149 | |
Insoluble in water. | |
[Zn++].[O-][O-] | |
97.38 | |
97.38 | |
Odorless |
>150°C (decomposition) | |
Powder | |
O2Zn | |
UN1516 | |
zinc peroxide, zinc superoxide, zinc dioxide, zinc peroxide un1516 oxidizer, zinc peroxide, medicinal, unii-0i969dvm77, zinc peroxide, medicinal usp, zpo | |
IPTOGCUGCFHDSS-UHFFFAOYSA-N | |
zinc(2+) dioxidanediide | |
14809 | |
≈50% ZnO2 |
Chemical Identifiers
1314-22-3 | |
97.38 | |
IPTOGCUGCFHDSS-UHFFFAOYSA-N | |
14809 |
O2Zn | |
MFCD00137469 | |
zinc peroxide, zinc superoxide, zinc dioxide, zinc peroxide un1516 oxidizer, zinc peroxide, medicinal, unii-0i969dvm77, zinc peroxide, medicinal usp, zpo | |
[Zn++].[O-][O-] |
Safety and Handling
GHS H Statement
H272-H332-H315-H319-H335
May intensify fire
oxidizer.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c
H271
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: ZINC PEROXIDE
EINECSNumber : 215-226-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only