missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc perchlorate hexahydrate, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 4431530

 View more versions of this product

Catalog No. AA4431530



Specifications

Specifications

Zinc perchlorate hexahydrate
(metals basis)
Zn(ClO4)2·6H2O
UN1481
2clo4.zn.6h2o
PADPILQDYPIHQQ-UHFFFAOYSA-L
zinc;diperchlorate;hexahydrate
202253
Crystalline
2.25 g/mL
1.5
106°C
10025-64-6
Cl2H12O14Zn
MFCD00150370
14,10148
Soluble in ethyl alcohol and water.
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2]
372.362
372.37 (264.28 Anhydrous)
99.997%
Odorless
White
250g

Chemical Identifiers

10025-64-6
372.362
PADPILQDYPIHQQ-UHFFFAOYSA-L
202253
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2]
Cl2H12O14Zn
MFCD00150370
2clo4.zn.6h2o
zinc;diperchlorate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 237-122-0

RTECSNumber : SD0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents