Please login to your online account to display your discounted pricing

Zinc oxide, ZN-2225, 40% in 1,2-propanediol monomethyl ether acetate, NanoArc™, Alfa Aesar™

Chemical Identifiers

Name Note ZN-2225
CAS 1314-13-2
Physical Form Suspension
Assay Percent Range 40% in 1, 2-propanediol monomethyl ether acetate
Molecular Formula OZn
Formula Weight 81.37g/mol
MDL Number MFCD00011300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4563222

 
alfa aesar
45632-22
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4563236

 
alfa aesar
45632-36
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Ester-like
Color White
Quantity 100g
Name Note ZN-2225
CAS 1314-13-2
Physical Form Suspension
Assay Percent Range 40% in 1, 2-propanediol monomethyl ether acetate
Molecular Formula OZn
Formula Weight 81.37g/mol
MDL Number MFCD00011300