missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc oxide, NanoTek™ Z1102PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion w/ dispersant, Alfa Aesar™

Manufacturer:  Alfa Aesar 4492436

 View more versions of this product

Catalog No. AA4492436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Zinc oxide dispersion in Dowanol™ PMA suitable for incorporation in a wide variety of coatings formulations
Specifications

Specifications

Zinc oxide, NanoTek™ Z1102PMA
1314-13-2
OZn
UN1993
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
Ester-like
Z1102PMA
50% in 1, 2-propanediol monomethyl ether acetate
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Suspension
500g

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

GHS H Statement
H226

GHS P Statement
P210-P280-P240-P241-P233-P242-P243-P303+P361+P353-P403+P235-P501a

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents