missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Oxide USP Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 1314-13-2
Physical Form Powder
Molecular Formula ZnO
Formula Weight 81.39
Chemical Name or Material Zinc Oxide
Grade USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK8824212 Avantor Performance
882405
STAKMOR 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1314-13-2
ZnO
Zinc Oxide
Powder
81.39
USP

Specifications

1314-13-2
ZnO
STAKMOR
Zinc Oxide
Powder
81.39
2.5kg
USP