missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 0001 Orientation, Alfa Aesar™

0001 Orientation

$1,553.14 - $1,553.14

Chemical Identifiers

CAS 1314-13-2
Physical Form Substrate
Molecular Formula ZnO
Formula Weight 81.37
MDL Number MFCD00011109
Chemical Name or Material Zinc Oxide Substrate, Polished Two Sides
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45487KT SDS Alfa Aesar
45487KT
1 ea. Each for $1,553.14
Specifications Description & Specifications

Chemical Identifiers

1314-13-2
ZnO
MFCD00011109
Substrate
81.37
Zinc Oxide Substrate, Polished Two Sides

Specifications

1314-13-2
ZnO
MFCD00011109
White
Zinc Oxide Substrate, Polished Two Sides
Substrate
81.37
Odorless
1 ea.