missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc oxide substrate, 10x10x0.5mm, polished one side, 0001 orientation, Thermo Scientific Chemicals
0001 Orientation
Supplier: Thermo Scientific Chemicals 045486KT
Description
Zinc oxide substrate is one of the suitable substrate for use as wide band gap semiconductor application. It is also used in power device, LED, sensor and detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc Oxide Substrate | |
White | |
Substrate | |
OZn | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
oxozinc | |
14806 | |
81.37 |
Polished One Side | |
1314-13-2 | |
1 Ea. | |
MFCD00011300 | |
Insoluble in water and alcohol. Soluble in acids and alkalies. | |
O=[Zn] | |
81.38 | |
CHEBI:36560 | |
Odorless |
Safety and Handling
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 215-171-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only