missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 0001 Orientation, Alfa Aesar™

0001 Orientation

$925.11 - $925.11

Chemical Identifiers

CAS 1314-13-2
Physical Form Substrate
Molecular Formula ZnO
Formula Weight 81.37
MDL Number MFCD00011109
Chemical Name or Material Zinc Oxide Substrate, Polished One Side
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45486KT SDS Alfa Aesar
45486KT
1 ea. Each for $925.11
Specifications Description & Specifications

Chemical Identifiers

1314-13-2
ZnO
MFCD00011109
Substrate
81.37
Zinc Oxide Substrate, Polished One Side

Specifications

Substrate
1 ea.
MFCD00011109
1314-13-2
Zinc Oxide Substrate, Polished One Side
81.37
White
ZnO
Odorless