Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Zinc oxide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 81.37
MDL Number MFCD00011300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41124KS

  SDS
alfa aesar™
41124KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 81.37
MDL Number MFCD00011300
Assay Percent Notes (metals basis)
Quantity 1 each
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.9%
TSCA TSCA
Chemical Name or Material Zinc oxide sputtering target