Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.9% (metals basis)

Magnetron sputtering source

Chemical Identifiers

Chemical Name or Material Zinc oxide sputtering target
CAS 1314-13-2
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Linear Formula ZnO
Formula Weight 81.37
MDL Number MFCD00011300
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41123KS

 
alfa aesar™
41123KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 81.37
Chemical Name or Material Zinc oxide sputtering target
TSCA TSCA
Odor Odorless
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Physical Form Target
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula ZnO
MDL Number MFCD00011300
CAS 1314-13-2
UN Number UN3077
Color White to pale yellow
Melting Point 1975°C
Quantity 1 each
Density 5.606g/mL
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number ZH4810000