Please login to your online account to display your discounted pricing

Zinc Oxide (Powder/USP), Fisher Chemical


Quantity 2.5kg
Packaging Poly Pail

Manufacturer: fisher scientific company  FLZ250212

 View more versions of this product

Catalog No. Z50-212

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Melting Point 1975°C
Formula Weight 81.39
Quantity 2.5kg
Purity Grade USP
Assay 99.0 to 100.5% (USP)
pH 7
Density 5.600g/cm³
Color Off-white
Molecular Formula OZn
Residual Solvents Meets Requirements (USP)
Packaging Poly Pail
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
ChemAlert Storage Symbol Gray
Chemical Name or Material Zinc Oxide
CAS 1314-13-2
MDL Number MFCD00011300
Heavy Metals and Iron (as Pb) Pass Test (USP)
Alkalinity Pass Test (USP)
Loss on Ignition 1.0% max. (USP)
Arsenic (As) 6ppm max. (USP)
Lead (Pb) Pass Test (USP)
Identification Pass Test (USP)
Carbonate and Color of Solution (USP) Pass Test

Specifications

Chemical Name or Material Zinc Oxide
CAS 1314-13-2
Purity Grade USP
Physical Form Powder/Solid
Assay 99.0 to 100.5% (USP)
Molecular Formula OZn
Formula Weight 81.39
MDL Number MFCD00011300
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents