Please login to your online account to display your discounted pricing

Zinc Oxide (Powder/USP), Fisher Chemical


Quantity 12kg
Packaging Fiber Drum

Manufacturer: fisher scientific company  FLZ5012

 View more versions of this product

Catalog No. Z50-12

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 81.39
Melting Point 1975°C
Physical Form Powder/Solid
Quantity 12kg
Assay 99.0 to 100.5% (USP)
Purity Grade USP
Molecular Formula OZn
Color Off-white
pH 7
Density 5.600g/cm³
Alkalinity Pass Test (USP)
Residual Solvents Meets Requirements (USP)
Lead (Pb) Pass Test (USP)
CAS 1314-13-2
Chemical Name or Material Zinc Oxide
MDL Number MFCD00011300
Heavy Metals and Iron (as Pb) Pass Test (USP)
Packaging Fiber Drum
Identification Pass Test (USP)
Carbonate and Color of Solution (USP) Pass Test
Arsenic (As) 6ppm max. (USP)
Loss on Ignition 1.0% max. (USP)
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Health Hazard 2 WARNING!
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Zinc Oxide
CAS 1314-13-2
Purity Grade USP
Physical Form Powder/Solid
Assay 99.0 to 100.5% (USP)
Molecular Formula OZn
Formula Weight 81.39
MDL Number MFCD00011300
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents