Please login to your online account to display your discounted pricing

Zinc Oxide (Powder/USP), Fisher Chemical

$457.86 - $3,432.93

Chemical Identifiers

Chemical Name or Material Zinc Oxide
CAS 1314-13-2
Purity Grade USP
Physical Form Powder/Solid
Assay 99.0 to 100.5% (USP)
Molecular Formula OZn
Formula Weight 81.39
MDL Number MFCD00011300
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

Z50-212

  SDS
fisher scientific company
FLZ250212
Poly Pail 2.5kg Each for $457.86

Z50-12

  SDS
fisher scientific company
FLZ5012
Fiber Drum 12kg Each for $3,432.93
Description & Specifications

Specifications

Physical Form Powder/Solid
Formula Weight 81.39
Melting Point 1975°C
Quantity 2.5kg
Purity Grade USP
Assay 99.0 to 100.5% (USP)
Molecular Formula OZn
Color Off-white
pH 7
Density 5.600g/cm³
Packaging Poly Pail
Residual Solvents Meets Requirements (USP)
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Chemical Name or Material Zinc Oxide
MDL Number MFCD00011300
CAS 1314-13-2
ChemAlert Storage Symbol Gray
Heavy Metals and Iron (as Pb) Pass Test (USP)
Alkalinity Pass Test (USP)
Lead (Pb) Pass Test (USP)
Loss on Ignition 1.0% max. (USP)
Arsenic (As) 6ppm max. (USP)
Identification Pass Test (USP)
Carbonate and Color of Solution (USP) Pass Test
Safety and Handling
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

WARNING!