Please login to your online account to display your discounted pricing

Zinc Oxide (Powder/Certified ACS), Fisher Chemical


Quantity 10kg
Packaging Poly Pail

Manufacturer: fisher scientific company  Z5210

 View more versions of this product

Catalog No. Z52-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 81.41
Melting Point 1975°C
Quantity 10kg
Molecular Formula OZn
Viscosity [NULL]
Color Off-white
pH 7
Density 5.600g/cm³
Potassium (K) 0.01% max.
Iron (Fe) 0.001% max.
Sodium (Na) 0.05% max.
Chemical Name or Material Zinc Oxide
Alkalinity Pass Test
Insoluble in Sulfuric Acid 0.01% max.
Lead (Pb) 0.005% max.
Manganese (Mn) 5ppm max.
Nitrate 0.003% max.
Packaging Poly Pail
MDL Number MFCD00011300
ChemAlert Storage Symbol Gray
Calcium (Ca) 0.005% max.
Identification Pass Test
Magnesium (Mg) 0.005% max.
Assay Percent Range ≥99.0 %
Sulfur Compounds 0.01% max.
Chloride 0.001% max.
CAS 1314-13-2
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

Specifications

Chemical Name or Material Zinc Oxide
CAS 1314-13-2
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula OZn
Formula Weight 81.41
MDL Number MFCD00011300
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents