Please login to your online account to display your discounted pricing

Zinc Oxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$165.47 - $404.89

Chemical Identifiers

Chemical Name or Material Zinc Oxide
CAS 1314-13-2
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula ZnO
Formula Weight 81.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004605

 
avantor performance
435805
Poly Bottle 2kg Each for $404.89

02004606

 
avantor performance
435801
Poly Bottle 500g Each for $165.47
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 81.39
Chemical Name or Material Zinc Oxide
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 5.61kg
Molecular Formula ZnO
CAS 1314-13-2
Quantity 2kg