missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc oxide, NanoTek™ Z1102PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion w/ dispersant, Alfa Aesar™

Manufacturer:  Alfa Aesar 44924A3

 View more versions of this product

Catalog No. AA44924A3



Description

Description

  • Zinc oxide dispersion in Dowanol™ PMA suitable for incorporation in a wide variety of coatings formulations
Specifications

Specifications

Zinc oxide
1314-13-2
OZn
UN1993
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
50% in 1,2-propanediol monomethyl ether acetate
70nm APS for dry (typical)
NanoTek™ Z1102PMA, colloidal dispersion with dispersant
50% in 1, 2-propanediol monomethyl ether acetate
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Powder
Ester-like
2kg

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents