missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, NanoTek™ C1, 99%, organosilane coated, hydrophobic, nonpolar

Manufacturer:  Alfa Aesar™ 4490022

 View more versions of this product

Catalog No. AA4490022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • NanoTek™ zinc oxide C1 has a hydrophobic, nonpolar organosilane coating to aid dispersing in organic systems
Specifications

Specifications

Zinc oxide, NanoTek™ C1
1314-13-2
MFCD00011300
14,10147
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
99%
10 to 25m2/g
100g
Organosilane coated, Hydrophobic, Nonpolar
OZn
UN3077
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Powder
40 to 100nm APS
1975°C

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : ZH4810000

Documents