missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, NanoTek™ C1, 99%, organosilane coated, hydrophobic, nonpolar

$42.55 - $42.55

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.379
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4490022 View Documents Alfa Aesar™
4490022
100g Each for $42.55
 
AA4490036 View Documents Alfa Aesar™
4490036
500g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • NanoTek™ zinc oxide C1 has a hydrophobic, nonpolar organosilane coating to aid dispersing in organic systems
Specifications

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
akro-zinc bar 85
CHEBI:36560
O=[Zn]

Specifications

1314-13-2
MFCD00011300
14,10147
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
99%
10 to 25m2/g
100g
OZn
UN3077
akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Powder
40 to 100nm APS
1975°C
Zinc oxide, NanoTek™ C1, Organosilane coated, Hydrophobic, Nonpolar
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : ZH4810000