Learn More
Zinc oxide, NanoTek|r, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04489922
Description
NanoTek™ zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systemsZinc oxide nanoparticles are used in the manufacture of rubber and cigarettes. It plays an important role in paints as a coating agent. It is used in creams and ointments, which are used to treat skin diseases. It acts as an additive in the manufacture of concrete. It is also used in ceramic industry and as an additive in food products such as breakfast cereals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc Oxide | |
1314-13-2 | |
100 g | |
MFCD00011300 | |
Insoluble in water. Soluble in dilute acetic acid,mineral acids,ammonia,ammonium carbonate and fixed alkali hydroxide solution. | |
O=[Zn] | |
81.38 | |
CHEBI:36560 | |
5.6 |
1975°C | |
Powder | |
OZn | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
oxozinc | |
14806 | |
81.37 |
Chemical Identifiers
1314-13-2 | |
81.38 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
14806 | |
oxozinc |
OZn | |
MFCD00011300 | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
CHEBI:36560 | |
O=[Zn] |
Safety and Handling
This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
DOTInformation : DOT Class: 9; Packaging Group: III
EINECSNumber : 215-222-5
RTECSNumber : ZH4810000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only