missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, NanoShield™ ZN-3008C, 50% in H2O, colloidal dispersion with cationic dispersant

Manufacturer:  Alfa Aesar™ 4500936

 View more versions of this product

Catalog No. AA4500936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • NanoShield™ ZN-3008C is a cationic dispersion of zinc oxide in water designed to be incorporated in coatings applied to a fabric or non-woven substrates
  • Zinc oxide is generally used to provide UVa and UVb blocking
Specifications

Specifications

Zinc oxide, NanoShield™ ZN-3008C
1314-13-2
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Dry Powder (Typical)
Odorless
500g
Colloidal dispersion with cationic dispersant
OZn
UN3082
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
50% in H2O
70nm APS

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA

Documents