missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, nanopowder, 99+% (metals basis)

Manufacturer:  Alfa Aesar™ 4429909

 View more versions of this product

Catalog No. AA4429909



Specifications

Specifications

Zinc oxide
(metals basis)
MFCD00011300
14,10147
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
≥99%
Odorless
1975°C
1314-13-2
OZn
UN3077
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Nanopowder
5.606g/mL
7 to 13nm APS
10g

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : ZH4810000

TSCA : TSCA

SDS Documents