missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, nanopowder, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 4453318

 View more versions of this product

Catalog No. AA4453318



Specifications

Specifications

Zinc oxide
(metals basis)
MFCD00011300
14,10147
Insoluble in water.
O=[Zn]
81.379
CHEBI:36560
20-30nm APS Powder
5.606 g/mL
20 to 30nm APS
1975°C
1314-13-2
OZn
UN3077
akro-zinc bar 85
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
99%
Odorless
White
50g

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 215-222-5

RTECSNumber : ZH4810000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents