missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, NanoGard™, low lead, 99+%

Manufacturer:  Alfa Aesar™ 4501036

 View more versions of this product

Catalog No. AA4501036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • NanoGard™ zinc oxide provides effective UV attenuation while maintaining clarity in silky-smooth, non-sticky formulations
  • Certified to less than 10ppm lead
Specifications

Specifications

Zinc oxide NanoGard™
1314-13-2
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Powder
Odorless
500g
Low lead
OZn
UN3077
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
≥99%
40 to 100nm APS

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : ZH4810000

TSCA : TSCA

Documents