Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, NanoArc™ ZN-2656, 50% in H2O, colloidal dispersion with nonionic dispersant and rheology modifier

Chemical Identifiers

Chemical Name or Material Zinc oxide
CAS 1314-13-2
Name Note NanoArc™ ZN-2656, colloidal dispersion with nonionic dispersant and rheology modifier
Formula Weight 81.37
MDL Number MFCD00011300
UN Number UN3082
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4722222

  SDS
alfa aesar™
4722222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4722236

  SDS
alfa aesar™
4722236
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 81.37
Name Note NanoArc™ ZN-2656, colloidal dispersion with nonionic dispersant and rheology modifier
MDL Number MFCD00011300
Quantity 100g
UN Number UN3082
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Physical Form Powder
Odor Odorless
Concentration 50% in H2O
CAS 1314-13-2
Particle Size 70nm APS for dry (typical)
Chemical Name or Material Zinc oxide