missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxide, NanoArc™ ZN-2650, 50% in H2O, colloidal dispersion with nonionic dispersant

Manufacturer:  Alfa Aesar™ 4490536

 View more versions of this product

Catalog No. AA4490536



Description

Description

  • NanoShield™ ZN-3010 is a dispersion of zinc oxide in water designed to be incorporated in coatings applied to a fabric or non-woven substrates
  • Zinc oxide is generally used to provide UVa and UVb blocking
Specifications

Specifications

Zinc oxide, NanoArc™ ZN-2650
1314-13-2
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Dry Powder (Typical)
Odorless
White
Colloidal dispersion with nonionic dispersant
OZn
UN3082
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
50% in H2O
70nm APS
500g

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA

Documents