Please login to your online account to display your discounted pricing

Zinc oxide doped with silver, NanoTek™, Alfa Aesar™

Chemical Identifiers

Name Note doped with silver
CAS 1314-13-2
Physical Form Suspension
Assay Percent Range 50% in H2O
Molecular Formula ZnO doped with Ag
Formula Weight 81.37g/mol
MDL Number MFCD00011300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4541022

 
alfa aesar
45410-22
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4541036

 
alfa aesar
45410-36
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Color Pink
Quantity 100g
Name Note doped with silver
CAS 1314-13-2
Physical Form Suspension
Assay Percent Range 50% in H2O
Molecular Formula ZnO doped with Ag
Formula Weight 81.37g/mol
MDL Number MFCD00011300