missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Oxide, ACS,Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Manufacturer:  MilliporeSigma 1088490500

 View more versions of this product

Catalog No. M1088490500



Description

Description

For analysis
Specifications

Specifications

Zinc oxide
OZn
White solid
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
ACS/Reagent Ph. Eur.
Plastic bottle
1314-13-2
UN 3077
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Solid
Odorless
500g

Chemical Identifiers

1314-13-2
81.379
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
OZn
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
Safety and Handling

Safety and Handling

H410: Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-222-5

Documents
Promotions

Promotions