missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Oxide, ACS,Reag. Ph Eur, For Analysis, EMSURE™, EMD Millipore™

$251.17 - $5,007.23

Chemical Identifiers

CAS 1314-13-2
Molecular Formula ZnO
Formula Weight 81.37
UN Number UN 3077
Chemical Name or Material Zinc oxide
Grade ACS/Reagent Ph Eur
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
M1088490500 Emd Millipore Corporation
1088490500
Each for $251.17
M1088491000 Emd Millipore Corporation
1088491000
Each for $417.89
M1088499025 Emd Millipore Corporation
1088499025
Each for $5,007.23
Specifications

Description

For analysis
Description & Specifications

Chemical Identifiers

1314-13-2
81.37
Zinc oxide
ZnO
UN 3077
ACS/Reagent Ph Eur

Specifications

1314-13-2
ZnO
81.37
Odorless
Zinc oxide
Solid
White solid
UN 3077
Plastic bottle
ACS/Reagent Ph Eur

Safety and Handling

H410: Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-222-5