Please login to your online account to display your discounted pricing

Zinc oxide, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  422740050

 View more versions of this product

Catalog No. AC422740050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 81.38
Purity Grade ACS Reagent Grade
Physical Form Powder
Quantity 5g
Color White
Packaging Glass Bottle
Density 5.6000g/mL
Sulfur Compounds 0.01% max.
Insoluble Matter 0.01% max. (in dilute H2SO4)
Sodium (Na) 500ppm max.
Linear Formula ZnO
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Chemical Name or Material Zinc oxide
Molecular Formula OZn
MDL Number MFCD00011300
CAS 1314-13-2
Calcium (Ca) 50ppm max.
Nitrate 30ppm max.
Magnesium (Mg) 50ppm max.
Alkalinity Passes Test
Potassium (K) 100ppm max.
Iron (Fe) 10ppm max.
Lead (Pb) 50ppm max.
Chlorine (Cl) 10ppm max.
Manganese (Mn) 5ppm max.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Zinc oxide
CAS 1314-13-2
Purity Grade ACS Reagent Grade
Assay ≥99.0%
Molecular Formula OZn
Linear Formula ZnO
Formula Weight 81.38
MDL Number MFCD00011300
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents