missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc oxide, 99.9995% (metals basis), Puratronic™, Alfa Aesar™

Manufacturer:  Alfa Aesar 1285514

 View more versions of this product

Catalog No. AA1285514



Specifications

Specifications

1314-13-2
OZn
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Powder
White
99.9995% metal basis
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
Odorless
25g

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Documents