missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 315791000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
Zinc oxide | |
1314-13-2 | |
Powder | |
99.999% | |
ZnO | |
01,1294; 05,622 | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
oxozinc | |
14806 | |
81.38 | |
Trace Metal Basis | |
5.6 |
1975.0°C | |
White | |
100 g | |
OZn | |
MFCD00011300 | |
15, 10347 | |
Solubility in water: 1.6mg/l (29°C). Other solubilities: soluble in acids and alkalies,soluble in diluted acetic or mineral acids,ammoni,um carbonate,fixed alkali hydroxide solution,insoluble in alcohol | |
O=[Zn] | |
81.38 | |
CHEBI:36560 | |
99.999% | |
Glass bottle | |
5.6000g/mL |
Chemical Identifiers
1314-13-2 | |
81.38 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
14806 | |
oxozinc |
OZn | |
MFCD00011300 | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
CHEBI:36560 | |
O=[Zn] |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 215-222-5
RUO – Research Use Only