Learn More
Zinc oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01113709
Description
Zinc oxide is used as a sun screener. It serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. Zinc oxide plays an important role in textiles as UV-absorber. It can protect rubber from fungi and UV light. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc Oxide | |
1314-13-2 | |
10 g | |
OZn | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
oxozinc | |
14806 | |
81.37 |
1975°C | |
Powder | |
99.999%, metal basis | |
MFCD00011300 | |
Insoluble in water. Soluble in dilute acetic acid,and dilute mineral acids. | |
O=[Zn] | |
81.38 | |
CHEBI:36560 | |
5.606 |
Chemical Identifiers
1314-13-2 | |
81.38 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
14806 | |
oxozinc |
OZn | |
MFCD00011300 | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
CHEBI:36560 | |
O=[Zn] |
Safety and Handling
This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets
Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
DOTInformation : DOT Class: 9; Packaging Group: III
EINECSNumber : 215-222-5
RTECSNumber : ZH4810000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only