missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 08781218

 View more versions of this product

Catalog No. AA8781218



Description

Description

Zinc oxide is used as a sun screener. It serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. It plays an important role in textiles as a UV-absorber. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc oxide
1314-13-2
50 g
OZn
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
1975°C
Powder
99.99% (metals basis)
MFCD00011300
Insoluble in water. Soluble in dilute acetic acid,mineral acids,ammonia,ammonium carbonate and fixed alkalihydroxide solution.
O=[Zn]
81.38
CHEBI:36560
5.606 g/mL

Chemical Identifiers

1314-13-2
81.38
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 215-222-5

RTECSNumber : ZH4810000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only