missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc oxide, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8781230

 View more versions of this product

Catalog No. AA8781230



Specifications

Specifications

Zinc oxide
99.99% (metals basis)
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
250g
1314-13-2
OZn
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.379
CHEBI:36560
Powder

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

EINECSNumber : 215-222-5

SDS Documents