missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Oxide, 99.9% (Metal Basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 44263A1

 View more versions of this product

Catalog No. AA44263A1



Specifications

Specifications

Zinc Oxide
99.9%, metal basis
MFCD00011300
Insoluble in water. Soluble in dilute acetic acid,mineral acids,ammonia,ammonium carbonate and fixed alkalihydroxide solution.
O=[Zn]
81.379
CHEBI:36560
-200 Mesh Powder
1975°C
1314-13-2
OZn
akro-zinc bar 85
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
5.606
1kg

Chemical Identifiers

1314-13-2
81.379
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
akro-zinc bar 85
CHEBI:36560
O=[Zn]
Safety and Handling

Safety and Handling

This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

DOTInformation : DOT Class: 9; Packaging Group: III

EINECSNumber : 215-222-5

RTECSNumber : ZH4810000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents