missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc oxalate dihydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1298514

 View more versions of this product

Catalog No. AA1298514



Specifications

Specifications

Zinc oxalate dihydrate
(metals basis)
ZnC2O4·2H2O
UN3288
zinc oxalate dihydrate, zinc oxalate, c2o4zn.2h2o
C(=O)(C(=O)O)O.O.O.[Zn]
191.44
189.42 (153.39 Anhydrous)
99.999%
3.28g/mL
25g
4255-07-6
C2H6O6Zn
MFCD00150611
14,10146
PLYRAVNBPDIEOH-UHFFFAOYSA-N
oxalic acid;zinc;dihydrate
91886451
Powder
Puratronic™
White

Chemical Identifiers

4255-07-6
191.44
PLYRAVNBPDIEOH-UHFFFAOYSA-N
91886451
C(=O)(C(=O)O)O.O.O.[Zn]
C2H6O6Zn
MFCD00150611
zinc oxalate dihydrate, zinc oxalate, c2o4zn.2h2o
oxalic acid;zinc;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents