Learn More
Zinc oxalate dihydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01298522
Description
Zinc oxalate dihydrate is used as an chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc oxalate dihydrate | |
White | |
100 g | |
C2H4O6Zn | |
MFCD00150611 | |
14,10146 | |
Soluble in dilute mineral acids. | |
O.O.[Zn++].[O-]C(=O)C([O-])=O | |
189.43 | |
189.42 (153.39 Anhydrous) | |
Puratronic™ |
55906-21-3 | |
Powder | |
(metals basis) | |
ZnC2O4·2H2O | |
UN3288 | |
5-chloro-2-methoxyaniline hydrochloride, hindamine red r, azobase cas, hindasol red rc, stabamine red rc, devol red j, fast red r base, red base ciba i, red base irga i, red salt ciba i | |
WCKIDCVWRJUPFY-UHFFFAOYSA-L | |
5-chloro-2-methoxyaniline;hydrochloride | |
77957 | |
99.999% | |
3.28 g/mL |
Chemical Identifiers
55906-21-3 | |
MFCD00150611 |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 208-934-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only