missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1113618

 View more versions of this product

Catalog No. AA1113618



Specifications

Specifications

Zinc nitrate hexahydrate
(metals basis)
Zn(NO3)2·6H2O
UN1514
acmc-1bzdd
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Zn+2]
297.478
297.47 (189.38 Anhydrous)
99.998%
2.07g/mL
Hygroscopic
~36°C
10196-18-6
H12N2O12Zn
MFCD00149889
14,10144
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc;dinitrate;hexahydrate
15865313
Crystalline Aggregates
Puratronic™
Odorless
White
50g

Chemical Identifiers

10196-18-6
297.478
JGPSMWXKRPZZRG-UHFFFAOYSA-N
15865313
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Zn+2]
H12N2O12Zn
MFCD00149889
acmc-1bzdd
zinc;dinitrate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: ZINC NITRATE

RTECSNumber : ZH4775000

TSCA : TSCA

Documents