Please login to your online account to display your discounted pricing

Zinc Nitrate, 6-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$166.98 - $534.05

Chemical Identifiers

Chemical Name or Material Zinc Nitrate
CAS 10196-18-6
Name Note 6-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Zn(NO3)2·6H2O
Formula Weight 297.47
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004603

 
avantor performance
434405
Poly Bottle 2.5kg Each for $534.05

02004604

 
avantor performance
434401
Poly Bottle 500g Each for $166.98
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 297.47
CAS 10196-18-6
Chemical Name or Material Zinc Nitrate
Name Note 6-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Quantity 2.5kg
Packaging Poly Bottle
Specific Gravity 1L = 2.07kg
Molecular Formula Zn(NO3)2·6H2O