Please login to your online account to display your discounted pricing

Zinc Nitrate, 6-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  434401

 View more versions of this product

Catalog No. 02004604

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 297.47
CAS 10196-18-6
Molecular Formula Zn(NO3)2·6H2O
Name Note 6-Hydrate
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Quantity 500g
Specific Gravity 1L = 2.07kg
Chemical Name or Material Zinc Nitrate

Specifications

Chemical Name or Material Zinc Nitrate
CAS 10196-18-6
Name Note 6-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Zn(NO3)2·6H2O
Formula Weight 297.47