missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc naphthenate, ^=67% in mineral spirits (10% Zn), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03955436

 View more versions of this product

Catalog No. AA3955436



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc naphthenate
Colorless
>37.8° (100°F)
500 g
Liquid
12001-85-3
1.01 g/mL
MFCD00147699
UN1993

Chemical Identifiers

12001-85-3
MFCD00147699
Safety and Handling

Safety and Handling

GHS H Statement
H350-H304-H226-H315-H319-H335
May cause cancer.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only