Please login to your online account to display your discounted pricing

Zinc (Mossy/Technical, Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  Z2500

 View more versions of this product

Catalog No. Z2-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Melting Point 419°C
Formula Weight 65.41
Boiling Point 907°C
Quantity 500g
Physical Form Solid
Color Light Blue
Atomic Weight 65.37
Density 7.14g/cm³
Vapor Pressure 1mmHg at 487°C
Identification Pass Test
Packaging Poly Bottle
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Molecular Formula Zn
CAS 7440-66-6
MDL Number MFCD00011291
Additional Information Vapor Pressure: 1mmHg at 487°C
Health Hazard 2 CAUTION!
ChemAlert Storage Symbol Gray
Chemical Name or Material Zinc

Specifications

Chemical Name or Material Zinc
CAS 7440-66-6
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Zn
Formula Weight 65.41
MDL Number MFCD00011291
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents