Please login to your online account to display your discounted pricing

Zinc (Mossy/Technical, Fisher Chemical


Quantity 3kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  Z23

 View more versions of this product

Catalog No. Z2-3

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 907°C
Melting Point 419°C
Purity Grade Technical Grade
Formula Weight 65.41
Quantity 3kg
Physical Form Solid
Color Light Blue
Density 7.14g/cm³
Vapor Pressure 1mmHg at 487°C
Atomic Weight 65.37
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Packaging Poly Bottle
Health Hazard 2 CAUTION!
Chemical Name or Material Zinc
Molecular Formula Zn
ChemAlert Storage Symbol Gray
MDL Number MFCD00011291
Additional Information Vapor Pressure: 1mmHg at 487°C
CAS 7440-66-6
Identification Pass Test

Specifications

Chemical Name or Material Zinc
CAS 7440-66-6
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Zn
Formula Weight 65.41
MDL Number MFCD00011291
Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents