missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc (Mossy/Technical, Fisher Chemical

$274.23 - $1,114.54

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym blue powder
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Z2-500 SDS View Documents Fisher Chemical
Z2500
500g Poly Bottle Each for $274.23
Case of 6 Each for $1,645.38
 
Z2-3 SDS View Documents Fisher Chemical
Z23
3kg Poly Bottle Each for $1,114.54
Case of 4 Each for $4,458.16
 
Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
blue powder
CHEBI:30185
[Zn]

Specifications

Vapor Pressure: 1mmHg at 487°C
7440-66-6
Pass Test
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
Poly Bottle
CHEBI:30185
Solid
1mmHg at 487°C
Blue
907°C
500g
7.14g/cm³
Zn
blue powder
[Zn]
65.38
23994
65.41
Technical
65.37
419°C
Zinc
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0