Please login to your online account to display your discounted pricing

Zinc, Mossy, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$217.36 - $768.91

Chemical Identifiers

Chemical Name or Material Zinc
CAS 7440-66-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Zn
Formula Weight 65.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004601

 
avantor performance
426005
Poly Bottle 2.5kg Each for $768.91

02004602

 
avantor performance
426001
Poly Bottle 500g Each for $217.36
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 65.39
Chemical Name or Material Zinc
Physical Form Mossy
CAS 7440-66-6
Molecular Formula Zn
Packaging Poly Bottle
Quantity 2.5kg
Specific Gravity 1L = 7.14kg