Please login to your online account to display your discounted pricing

Zinc Metal AR Dust, Macron Fine Chemicals™


macron

Quantity 500g
Packaging Glass

Manufacturer: avantor performance  868104

 View more versions of this product

Catalog No. MK8681500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Glass
Quantity 500g
Chemical Name or Material Zinc Metal
CAS 7440-66-6
Physical Form Dust
Molecular Formula Zn
Formula Weight 65.39g/mol

Specifications

Chemical Name or Material Zinc Metal
CAS 7440-66-6
Physical Form Dust
Molecular Formula Zn
Formula Weight 65.39g/mol