Please login to your online account to display your discounted pricing

Zinc Metal AR Dust, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Zinc Metal
CAS 7440-66-6
Physical Form Dust
Molecular Formula Zn
Formula Weight 65.39
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK8681212

 
avantor performance
868106
Glass 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 65.39
Physical Form Dust
Chemical Name or Material Zinc Metal
Packaging Glass
CAS 7440-66-6
Molecular Formula Zn
Quantity 2.5kg