Zinc meso-tetraphenylporphine, Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals 03955202
 View more versions of this product
Catalog No. AA3955202
Description
Description
A porphyrin used in light emitting diodes. It has valuable biological applications in molecular biology, fluoroimmunoassay and new pharmaceutical developments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Zinc meso-tetraphenylporphine
Purple
C44H28N4Zn
0.5 g
Insoluble in water.
[Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
678.12
678.1
14074-80-7
Odorless
MFCD00012155,MFCD00012155
zinc meso-tetraphenylporphine
FGEZGTKFMCNEOZ-UHFFFAOYSA-N
5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc
91667917
Crystalline
Chemical Identifiers
14074-80-7
678.12
FGEZGTKFMCNEOZ-UHFFFAOYSA-N
91667917
[Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
C44H28N4Zn
MFCD00012155,MFCD00012155
zinc meso-tetraphenylporphine
5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc
Safety and Handling
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures