Please login to your online account to display your discounted pricing

Zinc Iodide, Ultra Dry, 99.995% (Metal Basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Zinc Iodide
Name Note Ampouled under argon
CAS 10139-47-6
Purity Grade Ultra dry
Physical Form Granular
Assay Percent Range 99.9%, metal basis
Molecular Formula ZnI2
Formula Weight 319.18
MDL Number MFCD00011299
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3572714

  SDS
alfa aesar
3572714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3572706

  SDS
alfa aesar
3572706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3572722

  SDS
alfa aesar
3572722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Zinc Iodide
Name Note Ampouled under argon
CAS 10139-47-6
Health Hazard 1 Causes burns. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
Health Hazard 2 Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
DOT Information DOT Class: 8
EINECS Number 233-396-0
Purity Grade Ultra dry
Physical Form Granular
Assay Percent Range 99.9%, metal basis
Molecular Formula ZnI2
Formula Weight 319.18
MDL Number MFCD00011299
Quantity 25g
Safety and Handling

Causes burns. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).