missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc(II) Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T129425G

 View more versions of this product

Catalog No. T129425G



Specifications

Specifications

Zinc(II) Trifluoromethanesulfonate
C2F6O6S2Zn
1759
CITILBVTAYEWKR-UHFFFAOYSA-L
zinc;trifluoromethanesulfonate
104671
Crystal-Powder at 20°C
White
54010-75-2
MFCD00013229
zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Zn+2]
363.506
363.51
≥98.0% (T)
25g

Chemical Identifiers

54010-75-2
363.506
CITILBVTAYEWKR-UHFFFAOYSA-L
104671
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Zn+2]
C2F6O6S2Zn
MFCD00013229
zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
zinc;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents